Order Entry
Export
ContactUsLinkComponent
Succinic acid ≥99% FCC
Succinic acid ≥99% FCC
Catalog # SPCMS1686-500GM
Supplier:  Spectrum Chemical
CAS Number:  
undefined
Succinic acid ≥99% FCC
Catalog # SPCMS1686-500GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    99% Min.

Specification Test Results

About this item

 :
 :118.09 g/mol
Merck Index:13,08953
Boiling Pt:235 °C (1013 hPa)
Melting Pt:182 °C
Density:1,56 g/cm³ (20 °C)
Flash Pt:206 °C
Storage Temperature:Ambient
EINECS:203-740-4
UN:3261
ADR:8,III
 :GHS07
CAS Number:110-15-6
Formula:HOOCCH₂CH₂COOH
MDL Number:MFCD00002789