Order Entry
Export
ContactUsLinkComponent
2-Butyne-1,4-diol ≥99%
Toxic
Irritant
Health
2-Butyne-1,4-diol ≥99%
Catalog # SPCMB2232-50KGBL
Supplier:  Spectrum Chemical
CAS Number:  
Toxic
Irritant
Health
2-Butyne-1,4-diol ≥99%
Catalog # SPCMB2232-50KGBL
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pk:
    50 kg
  • Seq:
    4418

Specifications

Specification Test Results

  • Assay:
    99% min.

Specification Test Results

About this item

 :
 :86.09 g/mol
Storage Temperature:Ambient
UN:2716
ADR:6.1,III
 :GHS06
 :GHS07
 :GHS08
CAS Number:110-65-6
Formula:C₄H₆O₂
MDL Number:MFCD00002915