Order Entry
Export
ContactUsLinkComponent
Potassium iodide ≥99.0%, granular FCC
Irritant
Health
Potassium iodide ≥99.0%, granular FCC
Supplier:  Spectrum Chemical
Certificates
Irritant
Health
Potassium iodide ≥99.0%, granular FCC
Supplier:  Spectrum Chemical

Specification Test Results

  • Assay (Dried Basis):
    99.0 - 101.5%
  • Iodate (IO₃):
    max. 4 mg/kg
  • Lead (Pb):
    max. 4 mg/kg
  • Loss On Drying:
    max. 1 %
  • Nitrate, Nitrite, and Ammonia:
    To Pass Test

Specification Test Results