Sodium benzoate ≥99.0%, powder FCC
Catalog # SPCMS1146-500GM
Supplier: Spectrum Chemical
CAS Number:
Specifications
- Pack type:Poly bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Assay (C₇H₅O₂Na; Anhydrous Basis):99.0 - 101.0%
- Alkalinity (As NaOH):max.0.04 %
- Lead:max. 2 mg/kg
- Water:max. 1.5 %
- Identification:To Pass Test
Specification Test Results
About this item
:
:144.11 g/mol
Storage Temperature:Ambient
CAS Number:532-32-1
Formula:C₆H₅COONa
MDL Number:MFCD00012463