


Formic acid FCC
:64-18-6
:
:HCO₂H
:MFCD00003297
:46.03 g/mol
:1,220
:1779
:8.3,II
:GHS05
:GHS07
:GHS02
:Ambient
- Assay (HCOOH):85.0% min.
- Dilution Test:To Pass Test
- Acetic Acid (CH₃COOH):max. 0.4 %
- Sulfate:max. 0.004 %
- Identification:To Pass Test