Order Entry
Export
Orders LinkContactUsLinkComponent
Capric acid ≥99%

Capric acid ≥99%

200 L
Catalog # SPCMD2659-200LTBL
Irritant
Supplier:   Spectrum Chemical
Capric acid ≥99%
Catalog # SPCMD2659-200LTBL
Supplier:  Spectrum Chemical
CAS Number:  334-48-5
Capric acid ≥99%

200 L

CAS Number:  334-48-5
Price
Inquire For Price
Log in for availability
1 * 200 L
Product Details & Documents

Decanoic Acid is also known as capric acid. It is a saturated fatty acid from coconut oil and palm kernel oil. It can also be found in the milk of several types of mammals.

Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Synonyms:  Decylic acid, Decanoic acid
  • CAS Number:334-48-5
  • Formula:C₁₀H₂₀O₂
  • MDL Number:MFCD00004441
  • Molecular Weight:172.27 g/mol
  • Merck Index:13,01764
  • Boiling Pt:268 °C (1013 hPa)
  • Melting Pt:29…32 °C
  • Density:0,893
  • Flash Pt:147 °C
  • Storage Temperature:Ambient
  • EINECS:206-376-4
Specifications

Specification Test Results

  • Assay:
    99% Min.

Specifications

  • Pk:
    200 L

Specifications