- Antibody type:Primary
- Antigen name:C3ORF31
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:101-200/316
- Form:Liquid
- Gene ID:132001
- Antigen synonyms:MMP37-like protein, mitochondrial|Chromosome 3 open reading frame 31|C3orf31|MMP37_HUMAN|MMP37 like protein mitochondrial|MMP37 like protein|MGC16471
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:33 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C3ORF31
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
C3orf31 (chromosome 3 open reading frame 31), also known as MGC16471 or DKFZp434E0519, is a 316 amino acid mitochondrial protein that belongs to the MMP37 family and may be involved in translocation of transit peptide-containing proteins across the mitochondrial inner membrane. C3orf24 is encoded by a gene that maps to human chromosome 3p25.2. Chromosome 3 is made up of approximately 214 million bases encoding over 1,100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumor suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome, osteogenesis imperfecta and Charcot-Marie-Tooth disease are a few of the numerous genetic diseases associated with chromosome 3.
Type: Primary
Antigen: C3ORF31
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 101-200/316
Host: Rabbit
Isotype: IgG
Reactivity: