Order Entry
Export
ContactUsLinkComponent
Anti-Claudin 19 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-Claudin 19 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-13749R-A647
Supplier:  Bioss
Anti-Claudin 19 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-13749R-A647
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Claudin 19
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 647
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Epitope:
    21-120/224
  • Form:
    Liquid
  • Gene ID:
    149461
  • Antigen synonyms:
    Claudin-19|CLDN19|HOMG5
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    23 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Claudin 19
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity.

Type: Primary
Antigen: Claudin 19
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 21-120/224
Host: Rabbit
Isotype: IgG
Reactivity: Mouse