Anti-Claudin 20 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-13748R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Claudin 20
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:41-140/219
- Form:Liquid
- Antigen synonyms:CLD20_HUMAN|CLDN20|Claudin-20
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:24 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Claudin 20
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity.
Type: Primary
Antigen: Claudin 20
Clonality: Polyclonal
Clone:
Conjugation: Cy5
Public Immunogen Range: 41-140/219
Host: Rabbit
Isotype: IgG
Reactivity: