- Antibody type:Primary
- Antigen name:Cadherin 20
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:301-400/801
- Form:Liquid
- Antigen synonyms:FLJ37047|CDH7L3|Cadherin 20, type 2|Cdh7|Cadherin 20|cadherin-20
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:82 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Cadherin 20
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
The cadherins are a family of Ca2+-dependent adhesion molecules that function to mediate cell-cell binding critical to the maintenance of structure and morphogenesis. Cadherins each contain a large extracellular domain at the N-terminus, which is characterized by a series of five homologous repeats, the most distal of which is thought to be responsible for binding specificity. Cadherin-20, also known as protocadherin gamma B4, is a member of the protocadherin gamma gene cluster. Expressed primariy in fibroblasts, cadherin-20 plays a critical role in neuronal cell-cell connections in the brain. Through its ability to mediate cell-cell adhesion, caderin-20 is thought to help the body respond to injury by regulating post-traumatic wound healing.
Type: Primary
Antigen: Cadherin 20
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/801
Host: Rabbit
Isotype: IgG
Reactivity: