Order Entry
Export
ContactUsLinkComponent
 
Anti-G protein alpha 16 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-13248R-A750
 :  Bioss
Anti-G protein alpha 16 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-13248R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    G protein alpha 16
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    301-374/374
  • Form:
    Liquid
  • Gene ID:
    2769
  • Antigen synonyms:
    GNA 16|Gq class|Guanine nucleotide binding protein alpha 15 subunit|G alpha 15|Guanine nucleotide-binding protein subunit alpha-16|G alpha 16|GNA15|G alpha-16|G-protein subunit alpha-15|Guanine nucleotide-binding protein subunit alpha-15|GNA 15|GNA15_HUMAN|GNA16|G-protein subunit alpha-16|Guanine nucleotide binding protein alpha 15|G alpha-15
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
  • Molecular weight:
    44 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human G protein alpha 16
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

Heterotrimeric G proteins function to relay information from cell surface receptors to intracellular effectors (1). Each of a very broad range of receptors specifically detects an extracellular stimulus (a photon, pheromone, odorant, hormone or neurotransmitter) while the effectors (i.e., adenylyl cyclase), which act to generate one or more intracellular messengers, are less numerous. In mammals, G protein alpha, Beta and Gamma polypeptides are encoded by at least 16, 4 and 7 genes, respectively (2-5). Most interest in G proteins has been focused on their a subunits, since these proteins bind and hydrolyze GTP and most obviously regulate the activity of the best studied effectors. Four distinct classes of G alpha subunits have been identified; these include Gs, Gi, Gq and Ga 12/13 (3,4). The Gi class comprises all the known a subunits that are susceptible to pertussis toxin modifications, including Ga i-1, Ga i-2, Ga i-3, Ga o, Ga t1, Ga t2, Ga z and Ga gust (4). Of these, the three Ga i subtypes function to open atrial potassium channels (6). Ga 16 is a member of the Gq subfamily and is expressed specifically in hematopoietic cells (7).

Type: Primary
Antigen: G protein alpha 16
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-374/374
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat