Fatty Acid Screening Library
Catalog # CAYM10504-50
Supplier: Cayman Chemical
CAS Number:
Specifications
- Pk:50 µl
- Description:Fatty acids with diverse biological activities in a 96-well matrix tube rack format
Specifications
About this item
For screening of variety of fatty acids.
- Contains >75 compounds
- Includes arachidonic, linoleic, eicosapentaenoic, stearic, α- and γ-linolenic, oleic, and palmitic acids
The Fatty Acid Screening Library contains more than 75 compounds of fatty acids with diverse biological activities in a 96-