Anti-Proteasome 20S beta 3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-9357R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 20S beta 3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:121-205/205
- Form:Liquid
- Gene ID:5684
- Antigen synonyms:proteasome prosome, macropain subunit, beta type 3|Proteasome chain 13|HC10 II|proteasome component C10 II|C10 II|Proteasome beta 3 subunit|20S Proteasome _3|Proteasome theta chain|PSMB3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Proteasome 20S beta 3
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The proteasome is a multicatalytic proteinase complex with a highly ordered ring shaped 20S core structure. The core structure is composed of 4 rings of 28 non identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin dependent process in a non lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides.
Type: Primary
Antigen: Proteasome 20S beta 3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 121-205/205
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat