Specifications
- Antibody type:Primary
- Antigen name:Proteasome 20S C2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:1-110/263
- Form:Liquid
- Gene ID:5682
- Antigen synonyms:PSC 2|PROS-30|psmA1|HC 2|PSMA 1|PSC2|Proteasome prosome macropain subunit alpha type 1|Proteasome alpha 1 subunit|Proteasome subunit nu|Macropain subunit nu|Proteasome component C2|PROS30|HC2|Multicatalytic endopeptidase complex subunit C2|Protein P30 33K|Proteasome nu chain|Proteasome subunit alpha type-1|Proteasome subunit alpha type 1|NU antibody PROS 30|20S Proteasome 1|Proteasome subunit alpha type I|30 kDa prosomal protein|Macropain subunit C2|PSA1_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Proteasome 20S C2
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Ubiquitin-dependent proteolysis mediates selective destruction of various cell cycle regulators, transcription factors and tumor suppressors. In eukaryotic cells, selective breakdown of cellular proteins is ensured by their ubiquitination and subsequent degradation by the 26S proteasome. At specific stages of development, embryo- and tissue-specific components of the 26S proteasome form, facilitating proteolysis. 20S Proteasome ?, also designated macropain subunit C2 or PROS-30, is a prosomal protein involved in a non-lysosomal ATP/ubiquitin-dependent proteolytic pathway. The entire proteasome is composed of at least 15 non-identical subunits which form a highly-ordered ring-shaped structure.
Type: Primary
Antigen: Proteasome 20S C2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-110/263
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat