Order Entry
Export
ContactUsLinkComponent
Anti-Proteasome 19S S7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-Proteasome 19S S7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-9349R-A680
Supplier:  Bioss
Anti-Proteasome 19S S7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-9349R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proteasome 19S S7
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    51-150/433
  • Form:
    Liquid
  • Gene ID:
    5701
  • Antigen synonyms:
    Nbla10058|26S protease regulatory subunit 7|S7|Proteasome 26S subunit ATPase 2|26S proteasome AAA-ATPase subunit RPT1|Protein MSS1|MSS1|PSMC2
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    49 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Proteasome 19S S7
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

The 26S protease is involved in the ATP-dependent degradation of ubiquitinated proteins. The regulatory (or ATPase) complex confers ATP dependency and substrate specificity to the 26S complex. In case of HIV-1 infection, positive modulator of Tat-mediated transactivation.

Type: Primary
Antigen: Proteasome 19S S7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/433
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat