Anti-Proteasome 19S S3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-9347R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 19S S3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:201-300/534
- Form:Liquid
- Gene ID:5709
- Antigen synonyms:26S proteasome regulatory subunit S3|26S proteasome regulatory subunit RPN3|Proteasome subunit p58|PSMD3_HUMAN|26S proteasome non-ATPase regulatory subunit 3|RPN3|PSMD3|TSTA2|Proteasome prosome, macropain 26S subunit non ATPase 3|26S proteasome non ATPase regulatory subunit 3|S3 antibody Tissue specic transplantation antigen 2|P58
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Proteasome 19S 10B
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Acts as a regulatory subunit of the 26 proteasome which is involved in the ATP-dependent degradation of ubiquitinated proteins.
Type: Primary
Antigen: Proteasome 19S S3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/534
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat