Specifications
- Antibody type:Primary
- Antigen name:Connexin-29
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:71-170/269
- Form:Liquid
- Antigen synonyms:connexin29|Connexin 31.3|Gap junction protein epsilon 1 29kDa|Gap junction protein gamma 3 30.2kDa|Gap junction protein epsilon 1|Connexin 29|CX23|connexin-29|Gap junction epsilon-1 protein |Cx31.3|Connexin-30.2|Gap junction gamma 3 protein|GJC 3 antibodyGJC3|Gap junction epsilon 1 protein|Cx29 |GJE 1|Gap junction protein gamma 3|Gap junction gamma-3 protein|CXG3_HUMAN|Cx30.2|Connexin-31.3|Connexin 30.2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:30 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from mouse Connexin 29
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Connexin 29 belongs to the connexin family and is a member of the epsilon-type subfamily. Connexin 29 is a membrane bound, multi-pass protein also known as gap junction epsilon-1 protein. A connexon, consisting of connexin hexamers, is a membrane bound structure that is integral in the formation of a gap junction. One gap junction consists of a cluster of closely packed pairs of transmembrane channels, the connexons, through which materials of low molecular weight diffuse from one cell to a neighboring cell. Connexin 29 expression is restricted to the central nervous system and is present in brain, spinal cord, and sciatic nerve samples. It has been suggested that connexin 29 in the mature CNS contributes minimally to gap junctional intercellular communication in oligodendrocyte cell bodies. Rather, connexin 29 is targeted to myelin where it, along with connexin 32, may contribute to connexin-mediated communication between adjacent layers of uncompacted myelin.
Type: Primary
Antigen: Connexin-29
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 71-170/269
Host: Rabbit
Isotype: IgG
Reactivity: