Order Entry
Export
ContactUsLinkComponent
Anti-DSCC1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-DSCC1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-7720R-A680
Supplier:  Bioss
Anti-DSCC1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-7720R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DSCC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    301-393/393
  • Form:
    Liquid
  • Gene ID:
    79075
  • Antigen synonyms:
    Defective in sister chromatid cohesion protein 1 homolog|Sister chromatid cohesion protein DCC1|MGC5528|DCC1_HUMAN|hDCC1|DSCC1|DCC1|Defective in sister chromatid cohesion 1 homolog
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    45 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human DSCC1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Loads PCNA onto primed templates regulating velocity, spacing and restart activity of replication forks. May couple DNA replication to sister chromatid cohesion through regulation of the acetylation of the cohesin subunit SMC3.

Type: Primary
Antigen: DSCC1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-393/393
Host: Rabbit
Isotype: IgG
Reactivity: Others