Order Entry
Export
ContactUsLinkComponent
Anti-PDCD7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-PDCD7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6834R-A750
Supplier:  Bioss
Anti-PDCD7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6834R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PDCD7
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Epitope:
    301-400/485
  • Form:
    Liquid
  • Gene ID:
    10081
  • Antigen synonyms:
    PDCD7_HUMAN|Pdcd7|HES18|Programmed cell death 7|MGC22015|Apoptosis-related protein ES18|Programmed cell death protein 7|ES18
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    55 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human PDCD7
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

This gene encodes a 59 kDa protein that is associated with the U11 small nuclear ribonucleoprotein (snRNP), which is a component of the minor U12-type spliceosome responsible for catalyzing pre-mRNA splicing of U12-type introns.

Type: Primary
Antigen: PDCD7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-400/485
Host: Rabbit
Isotype: IgG
Reactivity: Mouse