Order Entry
Export
ContactUsLinkComponent
 
Anti-MAGEA5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-6820R-A680
 :  Bioss
Anti-MAGEA5 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-6820R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    MAGEA5
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    41-124/124
  • Form:
    Liquid
  • Gene ID:
    4104
  • Antigen synonyms:
    MAGA5_HUMAN|Cancer/testis antigen family 1 member 5|MAGE 5a antigen|MAGE 5b antigen|Melanoma antigen family A 5|MAGE 5 antigen|MGC129526|MAGE5|MAGEA4|CT1.5|Cancer/testis antigen 1.5|Melanoma associated antigen 5
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    13 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human MAGEA5
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

MAGEA5 is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. This MAGEA gene encodes a protein that is C-terminally truncated compared to other family members, and this gene can be alternatively interpreted to be a pseudogene.

Type: Primary
Antigen: MAGEA5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 41-124/124
Host: Rabbit
Isotype: IgG
Reactivity: Human