Order Entry
Export
ContactUsLinkComponent
Anti-FASTKD5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-FASTKD5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6776R-A750
Supplier:  Bioss
Anti-FASTKD5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6776R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    FASTKD5
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Epitope:
    501-600/762
  • Form:
    Liquid
  • Gene ID:
    380601
  • Antigen synonyms:
    Kiaa1792|Fastkd5|C78212|FAST kinase domain-containing protein 5|mKIAA1792
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    84 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from mouse FASTKD5
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Representing about 2% of human DNA, chromosome 20 consists of approximately 63 million bases and 600 genes. Chromosome 20 contains a region with numerous genes expressed in the epididymis which are thought important for seminal production and some viewed as potential targets for male contraception. The PRNP gene encoding the prion protein associated with spongiform encephalopathies, like Creutzfeldt-Jakob disease, is found on chromosome 20. Amyotrophic lateral sclerosis, spinal muscular atrophy, ring chromosome 20 epilepsy syndrome and Alagille syndrome are also associated with chromosome 20.

Type: Primary
Antigen: FASTKD5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-600/762
Host: Rabbit
Isotype: IgG
Reactivity: Human