Anti-Destrin 19 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6630R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Destrin 19
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:85-165/165
- Form:Liquid
- Gene ID:11034
- Antigen synonyms:corn1|Sid 23|DSN|Destrin|DEST_HUMAN|Dstn|sid23p|Actin depolymerizing factor|Actin-depolymerizing factor|ACTDP|Destrin actin depolymerizing factor|ADF
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:19 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Destrin 19/ADF
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Actin-depolymerising protein. Severs actin filaments (F-actin) and binds to actin monomers (G-actin). Acts in a pH-independent manner.
Type: Primary
Antigen: Destrin 19
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 85-165/165
Host: Rabbit
Isotype: IgG
Reactivity: