Anti-kir 6.1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-6468R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:kir 6.1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:61-160/424
- Form:Liquid
- Gene ID:3764
- Antigen synonyms:uKATP-1|ATP-sensitive inward rectifier potassium channel 8|KCNJ8|Potassium channel, inwardly rectifying subfamily J member 8|KIR6.1|Inward rectifier K(+) channel Kir6.1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:48 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human kir 6.1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This potassium channel is controlled by G proteins. Inward rectifier potassium channels are characterised by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. Can be blocked by external barium (By similarity).
Type: Primary
Antigen: kir 6.1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 61-160/424
Host: Rabbit
Isotype: IgG
Reactivity: Rat