Anti-Rho GTPase activating protein 29 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6098R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Rho GTPase activating protein 29
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:301-400/1261
- Form:Liquid
- Gene ID:9411
- Antigen synonyms:Rho GTPase-activating protein 29|ARHGAP29|PTPL1-associated RhoGAP protein 1|PARG1|Rho-type GTPase-activating protein 29
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:142 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PARG1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state. Has strong activity toward RHOA, and weaker activity toward RAC1 and CDC42. May act as a specific effector of RAP2A to regulate Rho. In concert with RASIP1, suppresses RhoA signaling and dampens ROCK and MYH9 activities in endothelial cells and plays an essential role in blood vessel tubulogenesis.
Type: Primary
Antigen: Rho GTPase activating protein 29
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/1261
Host: Rabbit
Isotype: IgG
Reactivity: Human