Order Entry
Export
ContactUsLinkComponent
 
Anti-Gene 21 protein/NPRL2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-6079R-A680
 :  Bioss
Anti-Gene 21 protein/NPRL2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
  BOSSBS-6079R-A680
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Gene 21 protein/NPRL2
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    221-320/380
  • Form:
    Liquid
  • Antigen synonyms:
    NPR like 2|Tumor suppressor candidate 4|TUSC 4|TUSC4 protein|NPR2 like|Gene 21 protein|NPRL 2|NPR 2L|TUSC4|NPR L2|Homologous to yeast nitrogen permease candidate tumor suppressor|NPRL2_HUMAN|Homologous to yeast nitrogen permease|G21 protein|TUSC 4 protein|NPRL2
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    44 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Gene 21 protein/NPRL2
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

NPR2L is homologous to yeast nitrogen permease and is a candidate tumor suppressor, being a negative regulator of cell cycle. Most abundant in skeletal muscle, followed by brain, liver, and pancreas, with lower amounts in lung, kidney, placenta, and heart. Expressed in most lung cancer cell lines tested. There are two isoforms, produced by alternative splicing.

Type: Primary
Antigen: Gene 21 protein/NPRL2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 221-320/380
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat