Order Entry
Export
ContactUsLinkComponent
Anti-Annexin A9 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-Annexin A9 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5916R-A750
Supplier:  Bioss
Anti-Annexin A9 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-5916R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Annexin A9
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    261-345/345
  • Form:
    Liquid
  • Gene ID:
    8416
  • Antigen synonyms:
    ANXA9_HUMAN|annexin XXXI|pemphaxin|ANX31|ANXA 9|ANXA9|ANXA-9|annexin A9|annexin 31
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    38 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Annexin A9
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

ANXA9 may act as a low affinity receptor for acetylcholine and is expressed in the stratified squamous skin epithelium, but not in epithelia of other types (at protein level). ANXA9 is one of the target molecules recognised by auto in patients with pemphigus vulgaris, a rare, autoimmune skin disease in which epidermal blisters occur as the result of the loss of cell-cell adhesion.

Type: Primary
Antigen: Annexin A9
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 261-345/345
Host: Rabbit
Isotype: IgG
Reactivity: