Anti-BZW1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-12903R-A750
: Bioss
- Antibody type:Primary
- Antigen name:BZW1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:11-120/419
- Form:Liquid
- Antigen synonyms:Nbla10236|basic leucine-zipper protein BZAP45|BZAP45|bzw1|BZW1_HUMAN|KIAA0005|Basic leucine zipper and W2 domain-containing protein 1|Protein Orf|putative protein product of Nbla10236|Basic leucine zipper and W2 domain containing protein 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:48 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human BZW1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Enhances histone H4 gene transcription but does not seem to bind DNA directly.
Type: Primary
Antigen: BZW1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 11-120/419
Host: Rabbit
Isotype: IgG
Reactivity: Rat