Order Entry
Export
ContactUsLinkComponent
Anti-Blood Group Lewis a Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-Blood Group Lewis a Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-12870R-A680
Supplier:  Bioss
Anti-Blood Group Lewis a Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-12870R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Blood Group Lewis a
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    201-300/361
  • Form:
    Liquid
  • Antigen synonyms:
    Lewis A antigen
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    42 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Blood Group Lewis a
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Glycosyltransferases that mediate the region and stereoselective transfer of sugars, such as the fucosyltransferases, determine cell surface-carbohydrate profiles, which is an essential interface for biological recognition processes. Fucosyltransferases catalyse the covalent association of fucose to different positional linkages in sugar acceptor molecules. The carbohydrate moieties generated and covalently attached to cell surfaces are necessary to ensure a surface contour that satisfies physiological roles, which are reliant on adhesion molecules such as Selectins. Hematopoietic lineages rely on Fucosyltransferases to confer a surface carbohydrate phenotype, which mediates proper cell adhesion molecule recruitment and cell trafficking. Blood Group Lewis a is a carbohydrate determinant carried on both glycolipids and glycoproteins.

Type: Primary
Antigen: Blood Group Lewis a
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/361
Host: Rabbit
Isotype: IgG
Reactivity: