Specifications
- Antibody type:Primary
- Antigen name:Trim22/Staf-50
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:231-330/498
- Form:Liquid
- Gene ID:10346
- Antigen synonyms:RING finger protein 94|RNF94|TTRIM22RIM22|TRI22_HUMAN|50 kDa stimulated trans acting factor|Stimulated Trans Acting Factor homolog of Mouse Rpt 1 gene|STAF50|Trim22|Stimulated trans acting factor 50 kDa|Tripartite mot containing protein 22|Staf 50|Tripartite binding mot 22
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:57 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Trim22
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The tripartite motif (TRIM) family of proteins are characterised by a conserved TRIM domain that includes a coiled-coil region, a B-box type zinc finger, one RING finger and three zinc-binding domains. Staf-50 (50 kDa-stimulated trans-acting factor), also known as TRIM22 (tripartite motif-containing 22), RNF94 or GPSTAF50, is a 498 amino acid cytoplasmic protein that belongs to the TRIM family and, characteristic of TRIM family members, contains one RING-type zinc finger, one B box-type zinc finger and one SPRY domain. Induced by IFN-å and IFN-, Staf-50 is strongly expressed in ovary, spleen, thymus and peripheral blood leukocytes where it is thought to mediate the antiviral effects of IFN proteins. Additionally, Staf-50 is present in leukemic cells, suggesting a role in cancer formation and metastasis. Staf-50 exists as two alternatively spliced isoforms which are encoded by a gene that maps to human chromosome 11.
Type: Primary
Antigen: Trim22/Staf-50
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 231-330/498
Host: Rabbit
Isotype: IgG
Reactivity: Human