Order Entry
Export
ContactUsLinkComponent
Anti-5HT3A Receptor/SR-3A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-5HT3A Receptor/SR-3A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-12051R-A680
Supplier:  Bioss
Anti-5HT3A Receptor/SR-3A Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-12051R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5HT3A Receptor/SR-3A
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Epitope:
    31-130/478
  • Form:
    Liquid
  • Gene ID:
    3359
  • Antigen synonyms:
    5-HT3A|5-hydroxytryptamine receptor 3A|HTR3|5HT3R|5-hydroxytryptamine receptor 3|5-HT-3|5-HT3R|HTR3A|Serotonin-gated ion channel receptor|Serotonin receptor 3A|5-HT3-A
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    53 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human 5HT3A Receptor/SR-3A
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor is a ligand-gated ion channel, which when activated causes fast, depolarizing responses in neurons. It is a cation-specific, but otherwise relatively nonselective, ion channel.

Type: Primary
Antigen: 5HT3A Receptor/SR-3A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 31-130/478
Host: Rabbit
Isotype: IgG
Reactivity: