Anti-5HT2A Receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-12049R-A750
: Bioss
- Antibody type:Primary
- Antigen name:5HT2A Receptor
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:51-150/471
- Form:Liquid
- Gene ID:3356
- Antigen synonyms:5-HT-2|5-HT-2A|5 HT 2A|Serotonin receptor 2A|5 hydroxytryptamine receptor 2A|5 HT 2|HTR2A|HTR 2A|5-hydroxytryptamine receptor 2A|5-hydroxytryptamine 2A receptor|5HT2A_HUMAN|5 HT2 receptor|HTR 2|HTR2|5 HT2A|SR-2A
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:53 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human 5HT2A Receptor
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system. This receptor is involved in tracheal smooth muscle contraction, bronchoconstriction, and control of aldosterone production.
Type: Primary
Antigen: 5HT2A Receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/471
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat