- Antibody type:Primary
- Antigen name:MTMR2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:371-450/643
- Form:Liquid
- Antigen synonyms:MTMR2_HUMAN|Myotubularin-related protein 2|Myotubularin related protein 2|KIAA1073|MTMR2|CMT4B1|CMT4B
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:73 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MTMR2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Myotubularin and the myotubularin-related proteins (MTMR1-9) belong to a highly conserved family of eukaryotic phosphatases. They are protein tyrosine phosphatases that utilize inositol phospholipids, rather than phosphoproteins, as substrates. MTMR family members hydrolyse both Phosphatidylinositol 3-phosphate (PtdIns3P) and PtdInsP2. MTMR2 interacts with MTMR5, an inactive family member that increases the enzymatic activity of MTMR2 and dictates its subcellular localisation. Mutations in MTMR2 cause autosomal recessive Charcot-Marie-Tooth type 4B1 (CMT4B1), which is characterised by reduced nerve conduction velocities, focally folded myelin sheaths and demyelination. MTMR3 and MTMR4 can either interact with each other or self associate. MTMR6 regulates the activity of the calcium-activated potassium channel 3.1. MTMR9 regulates the activity of MTMR7 and MTMR8.
Type: Primary
Antigen: MTMR2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 371-450/643
Host: Rabbit
Isotype: IgG
Reactivity: