Anti-PCP4/PEP-19 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-11326R-A750
: Bioss
- Antibody type:Primary
- Antigen name:PCP4/PEP-19
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Epitope:21-62/62
- Form:Liquid
- Gene ID:5121
- Antigen synonyms:PEP-19|PCP4|Purkinje cell protein 4|Brain-specific antigen PCP-4|PEP19|Brain-specific polypeptide PEP-19
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:7 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PCP4
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Probable regulator of calmodulin Signalling.
Type: Primary
Antigen: PCP4/PEP-19
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-62/62
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat