Specifications
- Antibody type:Primary
- Antigen name:B3GALNT1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:231-331/331
- Form:Liquid
- Gene ID:8706
- Antigen synonyms:Globoside synthase|GLOB|P antibody P antigen synthase|3-N-acetylgalactosaminyltransferase|P blood group globoside|Galactosylgalactosylglucosylceramide beta-D-acetyl-galactosaminyltransferase|B3galnt1|b3Gal T3|Galactosylgalactosylglucosylceramide beta D acetyl galactosaminyltransferase|brainiac1|beta3Gal T3|Gb4Cer|Beta-1|UDP Gal:betaGlcNAc beta 1,3 galactosyltransferase, polypeptide 3 Globoside blood group|UDP-GalNAc:beta-1|3-GalNAc-T1|Beta3Gal-T3|UDP GalNAc:betaGlcNAc beta 1,3 galactosaminyltransferase, polypeptide 1 Globoside blood group|b3Gal-T3|beta 1,3 N acetylgalactosaminyltransferase 1 globoside blood group|UDP GalNAc:beta 1,3 N acetylgalactosaminyltransferase 1|galT3|Beta 1,3 GalTase 3|3-GalTase 3|B3GL1_HUMAN|Beta-3-Gx-T3|3-N-acetylgalactosaminyltransferase 1|UDP-N-acetylgalactosamine:globotriaosylceramide beta-1|Beta 3 Gx T3|B3GALT3|Beta 3 GalNAc T1|UDP N acetylgalactosamine:globotriaosylceramide beta 1,3 N acetylgalactosaminyltransferase|Beta3GalT3|Beta 1,3 galactosyltransferase 3|P1 antibody UDP Gal:betaGlcNAc beta 1,3 galactosyltransferase 1|3-galactosyltransferase 3|GLCT3|globotriaosylceramide 3 beta N acetylgalactosaminyltransferase
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:39 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human B3GALNT1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This gene is a member of the beta-1,3-galactosyltransferase (beta3GalT) gene family. This family encodes type II membrane-bound glycoproteins with diverse enzymatic functions using different donor substrates (UDP-galactose and UDP-N-acetylglucosamine) and different acceptor sugars (N-acetylglucosamine, galactose, N-acetylgalactosamine). The beta3GalT genes are distantly related to the Drosophila Brainiac gene and have the protein coding sequence contained in a single exon. The beta3GalT proteins also contain conserved sequences not found in the beta4GalT or alpha3GalT proteins. The carbohydrate chains synthesised by these enzymes are designated as type 1, whereas beta4GalT enzymes synthesize type 2 carbohydrate chains. The ratio of type 1:type 2 chains changes during embryogenesis. By sequence similarity, the beta3GalT genes fall into at least two groups: beta3GalT4 and 4 other beta3GalT genes (beta3GalT1-3, beta3GalT5). The encoded protein of this gene does not use N-acetylglucosamine as an acceptor sugar at all. Multiple transcript variants that are alternatively spliced in the 5' UTR have been described; they all encode the same protein.
Type: Primary
Antigen: B3GALNT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 231-331/331
Host: Rabbit
Isotype: IgG
Reactivity: