Order Entry
Export
ContactUsLinkComponent
Anti-beta Amyloid 1-28 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-beta Amyloid 1-28 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-10429R-A680
Supplier:  Bioss
Anti-beta Amyloid 1-28 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-10429R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    beta Amyloid 1-28
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Epitope:
    1-28/42
  • Form:
    Liquid
  • Gene ID:
    351
  • Antigen synonyms:
    Amyloid Beta A4 Protein Precursor|Amyloid of Aging and Alzheimer Disease|beta Amyloid1-28|APP|A4|Amyloid Beta|Beta APP|PreA4|CTFgamma|CVAP|ABETA|Amyloid B|Amyloid 1-28|Cerebral Vascular Amyloid Peptide|beta-amyloid peptide|APPI|AD1|A4_HUMAN|Alzheimers Disease Amyloid Protein|B Amyloid|ABPP|beta-Amyloid 1-28|A beta|Protease nexin II|PN II|AAA|PN2
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    4,3 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide of human beta-Amyloid (1-28)
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

The cerebral and vascular plaques associated with Alzheimer's disease are mainly composed of Amyloid beta peptides. beta Amyloid is derived from cleavage of the Amyloid precursor protein and varies in length from 39 to 43 amino acids. beta Amyloid [1-40], beta Amyloid [1-42], and beta Amyloid [1-43] peptides result from cleavage of Amyloid precursor protein after residues 40, 42, and 43, respectively. The cleavage takes place by gamma-secretase during the last Amyloid precursor protein processing step. beta Amyloid [1-40], beta Amyloid [1-42], and beta Amyloid [1-43] peptides are major constituents of the plaques and tangles that occur in Alzheimer's disease. beta Amyloid and peptides have been developed as tools for elucidating the biology of Alzheimer's disease.

Type: Primary
Antigen: beta Amyloid 1-28
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 1-28/42
Host: Rabbit
Isotype: IgG
Reactivity: