Anti-C1orf157 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-9801R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:C1orf157
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:1-100/128
- Form:Liquid
- Gene ID:284573
- Antigen synonyms:Hypothetical protein LOC84284|Probable UPF0334 kinase like protein C1orf57|RP4 678E16.2|Chromosome 1 open reading frame 57|Nucleoside triphosphatase, cancer related|Human cancer related NTPase|CA157_HUMAN|Novel DUF829 domain containing protein|RP4 659I19.2|HCR NTPase|Nucleoside triphosphatase C1orf57|Nucleoside triphosphate phosphohydrolase|FLJ11383
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human C1orf157
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
C1orf57 (Chromosome 1 open reading frame 57) belongs to the UPF0334 family. The function of C1orf57 is unknown.
Type: Primary
Antigen: C1orf157
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/128
Host: Rabbit
Isotype: IgG
Reactivity: Human