Order Entry
Export
ContactUsLinkComponent
Anti-CLLD6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-CLLD6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-9611R-A750
 :  Bioss
Anti-CLLD6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-9611R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    CLLD6
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    22-120/196
  • Form:
    Liquid
  • Antigen synonyms:
    Chromosome 13 open reading frame 1|CLL deletion region gene 6 protein|Chronic lymphocytic leukemia deletion region gene 6 protein|Hypothetical protein LOC57213|CLLD6|CLLD 6|SPRY7_HUMAN
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human CLLD6/C13orf1
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

CLLD6, is a 196 amino acid protein that contains one B30.2/SPRY domain. Expression of CLLD6 is found at highest levels in heart, skeletal muscle and testis, with lower levels found in thymus, peripheral blood leukocytes, lymph node and bone marrow. Existing as two alternatively spliced isoforms, CLLD6 maps to human chromosome 13, which comprises nearly 4% of human DNA and contains about 114 million base pairs and 400 genes. Key tumour suppressor genes on chromosome 13 include the breast cancer susceptibility gene, BRCA2, and the RB1 (retinoblastoma) gene. RB1 encodes a crucial tumour suppressor protein which, when defective, leads to malignant growth in the retina and has been implicated in a variety of other cancers. The gene SLITRK1, which is associated with Tourette syndrome, is on chromosome 13. As with most chromosomes, polysomy of part or all of chromosome 13 is deleterious to development and decreases the odds of survival.

Type: Primary
Antigen: CLLD6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 22-120/196
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat