Specifications
- Antibody type:Primary
- Antigen name:TSPAN5
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:21-130/268
- Form:Liquid
- Gene ID:10098
- Antigen synonyms:Tetraspan 5|Tetraspan TM4SF|TSPAN5|NET-4|Tetraspanin 5|TSPAN 5|Tetraspanin-5|Transmembrane 4 superfamily member 9|NET 4|TSN5_HUMAN|Tetraspanin5|Transmembrane 4 superfamily member 8|TM4SF9|Tspan-5|Tetraspan NET-4|Tetraspan NET 4|NET4
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human TSPAN5
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
NET-4, also known as TSPAN5 or TM4SF9, is a 268 amino acid multi-pass membrane protein that belongs to the tetraspanin family and is thought to play a role in signal transduction events related to cell development, activation, growth and motility. The gene encoding NET-4 maps to human chromosome 4, which encodes nearly 6% of the human genome and has the largest gene deserts (regions of the genome with no protein encoding genes) of all of the human chromosomes. Defects in some of the genes located on chromosome 4 are associated with Huntington's disease, Ellis-van Creveld syndrome, methylmalonic acidemia and polycystic kidney disease.
Type: Primary
Antigen: TSPAN5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-130/268
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat