Order Entry
Export
ContactUsLinkComponent
Anti-IL-17 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-IL-17 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2140R-A680
Supplier:  Bioss
Anti-IL-17 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2140R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    IL-17
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    35-130/155
  • Form:
    Liquid
  • Gene ID:
    3605
  • Antigen synonyms:
    IL-17|IL17|CTLA8|IL17A|IL-17A|CTLA-8|Interleukin-17A|Cytotoxic T-lymphocyte-associated antigen 8
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    15 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human IL-17
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Induces stromal cells to produce proinflammatory and hematopoietic cytokines. Enhances the surface expression of ICAM1/intracellular adhesion molecule 1 in fibroblasts.

Type: Primary
Antigen: IL-17
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 35-130/155
Host: Rabbit
Isotype: IgG
Reactivity: Human