Anti-MMP-16 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1856R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:MMP-16
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:501-607/607
- Form:Liquid
- Gene ID:4325
- Antigen synonyms:Membrane-type matrix metalloproteinase 3|MMP-16|MT-MMP 3|MMP16|Membrane-type-3 matrix metalloproteinase|MT-MMP2|MTMMP3|MMP-X2|MT-MMP3|MMPX2|MT3-MMP|C8orf57|Matrix metalloproteinase-16|MT3MMP
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:56 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MMP-16
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Endopeptidase that degrades various components of the extracellular matrix, such as collagen type III and fibronectin. Activates progelatinase A. Involved in the matrix remodelling of blood vessels. Isoform short cleaves fibronectin and also collagen type III, but at lower rate. It has no effect on type I, II, IV and V collagen. However, upon interaction with CSPG4, it may be involved in degradation and invasion of type I collagen by melanoma cells.
Type: Primary
Antigen: MMP-16
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 501-607/607
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse