Specifications
- Antibody type:Primary
- Antigen name:Annexin IV
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:210-319/319
- Form:Liquid
- Gene ID:307
- Antigen synonyms:proliferation inducing protein 28.|P33/41|ANXA 4|DKFZp686H02120|ANX4|ANX A4|Annexin A4|P32.5|ANX 4|Carbohydrate binding protein P33/P41|Placental anticoagulant protein II antibody PP4X|Annexin IV placental anticoagulant protein II|Endonexin I|35 beta calcimedin|LipocortinIV|Lipocortin IV
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:35 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Annexin IV
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The Annexins are a family of structurally similar proteins. Annexins bind to phospholipids and may be involved in regulation of membrane transport, membrane channel activity, and interaction of the cell membrane with the extracellular matrix. Annexin A4 (ANXA4) belongs to the annexin family of calcium dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane related events along exocytotic and endocytotic pathways. ANXA4 has 45 to 59% identity with other members of its family and shares a similar size and exon intron organization. Isolated from human placenta, ANXA4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANXA4 is almost exclusively expressed in epithelial cells.
Type: Primary
Antigen: Annexin IV
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 210-319/319
Host: Rabbit
Isotype: IgG
Reactivity: