Anti-CK1+5+10+14 Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
Catalog # BOSSBS-1494R-HRP
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CK1+5+10+14
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:1452
- Antigen synonyms:K10|Hair alpha protein|Keratin type I cytoskeletal 10|Keratin type I cytoskeletal 14|67 kDa cytokeratin|CK 10|Cytokeratin 1 + 5 + 10+ 14|Keratin 5|K14|Keratin type II cytoskeletal 1|CK 5|CK 1|58 kDa cytokeratin|Keratin type II cytoskeletal 5.|Keratin 10|Keratin 14|CK 14|Keratin 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:40-68 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptides derived from human CK1+5+10+14
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Cytokeratins are a subfamily of intermediate filament proteins and are characterised by a remarkable biochemical diversity, represented in epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa. The individual cytokeratin polypeptides are designated 1 to 20.
Type: Primary
Antigen: CK1+5+10+14
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: