Order Entry
Export
ContactUsLinkComponent
Anti-Connexin-32 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-Connexin-32 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-1376R-FITC
Supplier:  Bioss
Anti-Connexin-32 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-1376R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Connexin-32
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Epitope:
    201-283/283
  • Form:
    Liquid
  • Gene ID:
    2705
  • Antigen synonyms:
    GJB1|Gap junction beta-1 protein|CX32|GAP junction 28 kDa liver protein|CMTX1|CMTX|Connexin-32
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    32 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Connexin-32
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

One gap junction consists of a cluster of closely packed pairs of transmembrane channels, the connexons, through which materials of low MW diffuse from one cell to a neighboring cell.

Type: Primary
Antigen: Connexin-32
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 201-283/283
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat