Anti-Connexin-32 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-1376R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Connexin-32
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Epitope:201-283/283
- Form:Liquid
- Gene ID:2705
- Antigen synonyms:GJB1|Gap junction beta-1 protein|CX32|GAP junction 28 kDa liver protein|CMTX1|CMTX|Connexin-32
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:32 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Connexin-32
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
One gap junction consists of a cluster of closely packed pairs of transmembrane channels, the connexons, through which materials of low MW diffuse from one cell to a neighboring cell.
Type: Primary
Antigen: Connexin-32
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 647
Public Immunogen Range: 201-283/283
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat