Anti-IL-17 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1183R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:IL-17
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:85-150/150
- Form:Liquid
- Gene ID:301289
- Antigen synonyms:IL-17|IL17|CTLA8|IL17A|IL-17A|CTLA-8|Interleukin-17A|Cytotoxic T-lymphocyte-associated antigen 8
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:15 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from rat IL-17
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Induces stromal cells to produce proinflammatory and hematopoietic cytokines. Enhances the surface expression of ICAM1/intracellular adhesion molecule 1 in fibroblasts.
Type: Primary
Antigen: IL-17
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 85-150/150
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat