Anti-Connexin-40 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-1050R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Connexin-40
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Epitope:301-358/358
- Form:Liquid
- Gene ID:2702
- Antigen synonyms:GJA5|Connexin-40|Gap junction alpha-5 protein|CX40
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:40 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Connexin-40
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
One gap junction consists of a cluster of closely packed pairs of transmembrane channels, the connexons, through which materials of low MW diffuse from one cell to a neighboring cell.
Type: Primary
Antigen: Connexin-40
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 301-358/358
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog