Order Entry
Export
ContactUsLinkComponent
Anti-Connexin-40 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-Connexin-40 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1050R-A680
Supplier:  Bioss
Anti-Connexin-40 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-1050R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Connexin-40
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    301-358/358
  • Form:
    Liquid
  • Gene ID:
    2702
  • Antigen synonyms:
    GJA5|Connexin-40|Gap junction alpha-5 protein|CX40
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    40 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Connexin-40
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

One gap junction consists of a cluster of closely packed pairs of transmembrane channels, the connexons, through which materials of low MW diffuse from one cell to a neighboring cell.

Type: Primary
Antigen: Connexin-40
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-358/358
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog