Order Entry
Export
ContactUsLinkComponent
Anti-MMP-20 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-MMP-20 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0985R-A750
Supplier:  Bioss
Anti-MMP-20 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0985R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MMP-20
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    401-483/483
  • Form:
    Liquid
  • Gene ID:
    9313
  • Antigen synonyms:
    Enamelysin|MMP-20|MMP20|Enamel metalloproteinase|Matrix metalloproteinase-20|AI2A2
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    43 kDa
  • Storage temperature:
    Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human MMP20
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Degrades amelogenin, the major protein component of the enamel matrix and two of the macromolecules characterizing the cartilage extracellular matrix: aggrecan and the cartilage oligomeric matrix protein (COMP). May play a central role in tooth enamel formation.

Type: Primary
Antigen: MMP-20
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 401-483/483
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse