Anti-MMP-20 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0985R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:MMP-20
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:401-483/483
- Form:Liquid
- Gene ID:9313
- Antigen synonyms:Enamelysin|MMP-20|MMP20|Enamel metalloproteinase|Matrix metalloproteinase-20|AI2A2
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:43 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MMP20
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Degrades amelogenin, the major protein component of the enamel matrix and two of the macromolecules characterizing the cartilage extracellular matrix: aggrecan and the cartilage oligomeric matrix protein (COMP). May play a central role in tooth enamel formation.
Type: Primary
Antigen: MMP-20
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 401-483/483
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse