Anti-NPY2R Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0937R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:NPY2R
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:301-381/381
- Form:Liquid
- Gene ID:4887
- Antigen synonyms:Neuropeptide Y receptor type 2|Y2 receptor|NPY2-R|NPY-Y2 receptor|NPY2R
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:43 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human NPY2R
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Receptor for neuropeptide Y and peptide YY. The rank order of affinity of this receptor for pancreatic polypeptides is PYY > NPY > PYY (3-36) > NPY (2-36) > [Ile-31, Gln-34] PP > [Leu-31, Pro-34] NPY > PP, [Pro-34] PYY and NPY free acid.
Type: Primary
Antigen: NPY2R
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-381/381
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat